Learn More
Cytiva TE 22 Mini Tank Transfer Unit
Performs protein and nucleic acid transfers for Western, Southern, and Northern blots in less than one hour
Brand:  Cytiva TE22
Product Code. 12984840
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes: Four gel cassettes; eight foam sponges, 3mm thick; four foam sponges, 6mm thick; and 25 sheets of blotter paper
Description
Description
  • Transfers four mini-gels up to 9 × 10cm in size in about 1L of buffer
  • Built-in alumina-covered heat exchanger dissipates heat generated by tank electrotransfer; typical temperature increase: ≤5°C
  • 80cm of wire in seven parallel strands on each electrode panel delivers a uniform electrical field
  • Hinged, color-coded cassettes allow quick setup of transfers
  • Replacement electrode panel, sponges, and gel cassettes available separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN61010-1, CE 73/23/EEC (LV directive)
Four gel cassettes, eight 3mm thick Foam sponges, four 6mm thick Foam sponges and 25 sheets of blotter Paper, Transphor tank
33.81 oz.
5.5 in.
6.5 in.
100 V
500 mA
TE 22 Mini Tank Transfer Unit
Transfers four mini-gels up to 9 x 10cm
1000 mL
14 cm
16.5 cm
50 w