Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Cassettes
For use with TE62 Standard Transfer Tank with Cooling Chamber
Brand:  Hoefer™ TE24
 View more versions of this product
Product Code. 12004807
  • 137.00 € / Each
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessories and Replacement Parts for TE62
Specifications
Specifications
TE 22 Mini Tank Transfer Unit
Gel Cassette
1 cassette assembly, 2 Foam sponges 3mm thick and 1 Foam sponge 6mm thick
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products