missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Product Code. 15875418
Click to view available options
Quantity:
100 mL
10 x 1 mL
500 mL
Packaging:
Nalgene bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Use directly to resuspend and/or dilute purified DNA or RNA

Use directly to resuspend and/or dilute purified DNA or RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Physical Form Solution
Quantity 500 mL
Packaging Nalgene bottle
Grade Molecular Biology
Chemical Name or Material TE Buffer

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'