missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade, Thermo Scientific Chemicals

Use directly to re-suspend and/or dilute purified DNA or RNA

60.90 € - 96.40 €

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15746272
View Documents Promotion Details
Thermo Scientific
J75793.AE
100 mL Nalgene bottle
60.90 €
100mL
Please to purchase this item. Need a web account? Register with us today!
15684018
View Documents Promotion Details
Thermo Scientific
J75793.AP
500 mL Nalgene bottle
96.40 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Use directly to re-suspend and/or dilute purified DNA or RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1185-53-1
157.594
93573
C(C(CO)(CO)N)O.Cl
C4H12ClNO3
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride

Specifications

1185-53-1
8
C4H12ClNO3
C(C(CO)(CO)N)O.Cl
157.594
Molecular Biology
1X
Solution
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
93573
TE Buffer
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only