Learn More
TE Buffer, Tris-EDTA, 100X Solution, pH 8.0, Molecular Biology, Fisher BioReagents™
Brand: Fisher BioReagents BP1338-4
This item is not returnable.
View return policy
Description
TE Buffer, Tris-EDTA, 100X Solution, pH 8.0, Molecular Biology
Tris–EDTA (TE) is routinely used for suspending nucleic acid samples.Specifications
Tris-EDTA | |
77-86-1, 60-00-4, 7732-18-5 | |
Liquid | |
DNase free | |
PolyPac™ | |
Molecular Biology |
100X Solution | |
Undesignated | |
4 L | |
Filtered through a 0.2-micron filter. | |
Protease free |