missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, Tris-EDTA, 10X Solution, pH 7.6, Molecular Biology, Fisher BioReagents™

23.15 € - 47.90 €

Specifications

Chemical Name or Material Tris-EDTA (TE), 10X Solution
Name Note pH 7.6
CAS 77-86-1, 6381-92-6, 7647-01-0, 7732-18-5
Color Colorless
pH 7.4 to 8.1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10153393
View Documents
Fisher BioReagents
BP2475-100
100 mL Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10638593
View Documents
Fisher BioReagents
BP2475-500
500 mL Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10409123
View Documents
Fisher BioReagents
BP2475-1
1 L Nalgene Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

TE Buffer, Tris-EDTA, 10X Solution, pH 7.6, Molecular Biology

Tris–EDTA (TE) is used for suspending nucleic acid samples.
Specifications

Specifications

Tris-EDTA (TE), 10X Solution
77-86-1, 6381-92-6, 7647-01-0, 7732-18-5
7.4 to 8.1
DNase free
Protease free
Molecular Biology
pH 7.6
Colorless
Liquid
Filtered through a 5-micron filter and autoclaved.
TE
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0

Recommended Storage : RT

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.