missing translation for 'onlineSavingsMsg'
Learn More

TEA, 0.2M buffer soln., pH 7.0, Thermo Scientific™

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym Triethanolamine buffer solution
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol
SMILES C(CO)N(CCO)CCO
View More Specs

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

102-71-6
149.19
GSEJCLTVZPLZKY-UHFFFAOYSA-N
7618
2-[bis(2-hydroxyethyl)amino]ethanol
C6H15NO3
MFCD00002855
Triethanolamine buffer solution
CHEBI:28621
C(CO)N(CCO)CCO

Specifications

102-71-6
7.0
C6H15NO3
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]ethanol
7618
TEA buffer soln.
0.2 M
Liquid
Odorless
Triethanolamine buffer solution
C(CO)N(CCO)CCO
149.19
CHEBI:28621
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.