missing translation for 'onlineSavingsMsg'
Learn More
Tebbe reagent, 0.5M solution in toluene, AcroSeal™ Product Code.: 10657664

Tebbe reagent, 0.5M solution in toluene, AcroSeal™

Product Code. 10657664
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10657664

Brand: Thermo Scientific Acros 426061000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

CAS: 67719-69-1 | C13H18AlClTi

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination), Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

CAS 67719-69-1, 108-88-3
Color Purple to Red
Density 0.9270g/mL
Flash Point 4°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C13H18AlClTi
MDL Number MFCD00151575
Quantity 100 mL
Merck Index 15, 9228
Specific Gravity 0.927
Synonym Bis(cyclopentadienyl-micron-chloro(dimethylaluminum-micron-methylenetitanium
Formula Weight 284.62
Physical Form Solution
Chemical Name or Material Tebbe reagent, 0.5M solution in toluene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tebbe reagent, 0.5M solution in toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tebbe reagent, 0.5M solution in toluene, AcroSeal™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.