missing translation for 'onlineSavingsMsg'
Learn More

Technical Buffer pH 2, Mettler Toledo™
GREENER_CHOICE

Product Code. 12601306
Click to view available options
Quantity:
6 x 250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Traceable to the primary standards of reference materials selected by NIST.

  • pH 2
TRUSTED_SUSTAINABILITY

Specifications

Description Technical buffer pH 2.00, 6 x 250mL
pH 2
Quantity 6 x 250 mL
Solution Type Buffer
Packaging 6x 250 mL
compliance-icons
Product Identifier
  • Technical buffer pH 2.00
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Citric acid anhydrous, Hydrogen chlorideanhydrous
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'