missing translation for 'onlineSavingsMsg'
Learn More

Technical Buffer Solution pH 4.01, Mettler Toledo™
GREENER_CHOICE

Product Code. 12611306
Click to view available options
Quantity:
6 x 250 mL
Packaging:
6x 250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Calibrate pH sensors with reliable, all-around pH buffers. Mettler Toledo Technical pH Buffer Solution is available in handy bottles with no-fuss labels. Six-packs provide larger quantities for convenient, economical supplies.

pH electrodes must be calibrated regularly; at least once a day prior to measurements and after every cleaning, regeneration or long-term storage. The calibration adjusts slope and offset of an electrode to their true values. Since an electrode is characterized by both its slope and its offset, it is advisable to do a minimum of a two point calibration for reliable measurements and better precision. It is important is that the measurements are within the calibration range.

The internationally recognized pH scale is based on standard reference materials (SRM) selected by NIST (National Institute of Standards and Technology, USA). These buffer solutions are traceable to these primary standards. The quality inspection certificate, that is available for every bottle and sachet, guarantees stated values and traceability.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
Color Red
Certificate of Analysis Supplied
Density 1g/cm3
Description Technical buffer pH 4.01, 6 x 250mL
pH 4.01
Physical Form Liquid
Quantity 6 x 250 mL
Formulation Potassium hydrogen phthalate
Packaging 6x 250 mL
Traceability to NIST Traceable to NIST
Solubility Information Completely miscible in water
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'