missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Technical Buffer Solution pH 7, WTW™
Click to view available options
Quantity:
250 mL
1000 mL
Packaging:
1 Bottle
10 Bottles
25 Bottles
Description
- Precise and traceable to NIST (National Institute of Standards and Technology, USA in bulk size of 250 ml with built-in dosing vessel standard buffer
- Life up to 3 years (unopened)
- Thrifty use
- Easiest handling by means of dosing device
Specifications
Specifications
pH | 7 |
Quantity | 1000 mL |
Packaging | 10 Bottles |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'