Learn More
Texwipe™ TechniCloth™ Mop Cover
Fabricated from a hydroentangled, nonwoven blend of 55% cellulose and 45% polyester
129.00 € - 172.00 €
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
15732416
|
Texwipe™
TX627C |
37.5 x 67.5 cm |
172.00 €
Pack of 150 |
In Stock
Log in to see stock. |
|||||
15742416
|
Texwipe™
TX630 |
59 x 25 cm |
129.00 €
Pack of 150 |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
Description
- Fabric blend combines the highly absorbent properties of a natural fiber with the cleanliness and strength of a synthetic wiper
- With its low particle counts, low extractable levels and solvent compatibility, TechniCloth™ is ideal for general wiping and spill control in controlled environments
- Ideal for applications where high absorbency and low extractables are prime considerations
- Excellent for picking up aqueous spills
- Excellent wet and dry strength due to the polyester
- Highly absorbent due to the cellulose
- Compatible with most solvents
- Contains no chemical binders
- Low extractable levels
Specifications
Polyester Cellulose |