Learn More
Medline™ Technico™ DIGIT-032 ATC Sugar Pocket refractometer
Brand: Medline™ 8145.0320M
This item is not returnable.
View return policy
Specifications
Pocket Sugar Refractometer | |
0 to 32 Brix | |
DIGIT-032 ATC | |
1 Each |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
8145.0320M
File size: 429.8 kB
Product Content Correction
Product Title
Medline™ Technico™ DIGIT-032 ATC Sugar Pocket refractometer > Brix Range: 0 to 32%
Spot an opportunity for improvement?Share a Content Correction