missing translation for 'onlineSavingsMsg'
Learn More

Technico™ Pocket Refractometer

Brand:  Technico 8100.2862

 View more versions of this product

Product Code. 10553353

  • 177.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Specifications

Specifications

Pocket Refractometer
DIGIT-2862
Brix
180 g
28 to 62 Brix
Saturated salt solution at 29,6%
Measuring mid-range concentrations of sugars
55 x 80 x 205 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Technico™ Pocket Refractometer > Scale Range: 28 to 62% Brix

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.