Learn More
Gallenkamp Vacuum Oven with Digital Electronic PID Control
Gallenkamp Vacuum Oven is insulated with special grade glass wool to reduce the radiation heat loss to the minimum.
Brand:  Gallenkamp OVA031.XX3.5
Product Code. 13025703
  • 5495.00 € / Each
Estimated Shipment: 07-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Double walled chamber, outer chamber rectangular in shape made of thick mild steel sheets
  • The inner chamber is cylindrical in shape made of stainless steel sheets with provision for vacuum pump and the vacuum gauge is provided at the top of unit
  • The inner chamber accommodate one no. Stainless steel tray at adjustable height.
  • The oven is insulated with special grade glass wool to reduce the radiation heat loss to the minimum
  • The door is of stainless steel plate with a specially designed Mechanism having alignment
  • A perfect seal is obtained by a positive screw specially designed for tightening the lid
  • A glass window is provided in the centre for viewing without opening the door
  • A set of band heaters is wrapped around the vacuum chamber for fast response and maximum uniformity of heat
Specifications
Specifications
31 L
200°C
Vacuum Oven
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gallenkamp Vacuum Oven with Digital Electronic PID Control > Capacity: 31L