missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Teepol™ Multi-purpose Detergent

Contains a balanced blend of anionic and nonionic constituents. Azpack™ Teepol™ Multi-purpose Detergent is a high quality neutral detergent designed for general washing up and cleaning.

Brand:  Azpack™ 12356327

Product Code. 12356327

  • 39.00 € / 5 L

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High quality neutral detergent
  • General all-purpose cleaner
  • Supplied in a 5L plastic container with handle
  • Wide mouth for efficient pouring
Specifications

Specifications

Liquid Detergent
Anionic and nonionic constituents
General purpose cleaning and washing up
5 L
Liquid
Plastic container with handle
1 Ea.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Teepol Multi Purpose Detergent

Supplemental information
  • MIXTURE LIST-Contains : Sodium Dodecylbenzene Sulphonate, SodiumC12-C15 Alcohol Ether Sulphate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers