missing translation for 'onlineSavingsMsg'
Learn More
Telenzepine dihydrochloride, Tocris Bioscience™ Product Code.: 30001200

Telenzepine dihydrochloride, Tocris Bioscience™

Product Code. 30001200
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001200

Brand: Tocris Bioscience 1122/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective M1 antagonist

Selective high affinity muscarinic M1 receptor antagonist (Ki = 0.94 nM). Inhibits gastric acid secretion.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147416-96-4
Molecular Formula C19H24Cl2N4O2S
Molecular Weight (g/mol) 443.387
InChI Key JTQJFSQQHGPLOX-UHFFFAOYSA-N
Synonym telenzepine dihydrochloride, telenzepine hydrochloride, prestwick_649, 4,9-dihydro-3-methyl-4-4-methyl-1-piperazinyl acetyl-10h-thieno 3,4-b 1,5 benzodiazepin-10-one dihydrochloride, 4,9-dihydro-3-methyl-4-4-methyl-1, piperazinyl acetyl-10h-thieno 3,4-b 1,5 benzodiazepin-10-one dihydrochloride, 1-methyl-10-2-4-methylpiperazin-1-yl acetyl-5h-thieno 3,4-b 1,5 benzodiazepin-4-one dihydrochlorideShow MoreShow Less
PubChem CID 6419995
IUPAC Name 1-methyl-10-[2-(4-methylpiperazin-1-yl)acetyl]-5H-thieno[3,4-b][1,5]benzodiazepin-4-one;dihydrochloride
SMILES CC1=C2C(=CS1)C(=O)NC3=CC=CC=C3N2C(=O)CN4CCN(CC4)C.Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 443.39
Percent Purity >98%
Chemical Name or Material Telenzepine dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Telenzepine dihydrochloride, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.