missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Telescopic Rod

Telescopic Rod is a versatile sampling system with exchangeable tools for a wide range of applications.

Brand:  Buerkle™ 5355-0300

54.71 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.


 View more versions of this product

Product Code. 10040822

  • 82.90 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Different tools can be attached to the telescopic rod
  • The tools (angular beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net) are attached to the telescopic rod by a practical snap-in joint
  • Ideal for sampling from baths, tanks, shafts, open water courses etc
  • Sampling depths down to 6 m can be reached with the adjustable telescopic aluminum rod
  • The telescopic rods cannot be rotated radially, which means even better handling
  • The individual rods have been provided with an end stop, this means that one rod cannot accidentally slip out of another rod
  • Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm. Fits into every car boot!
  • Springs made of stainless steel
Specifications

Specifications

Telescopic Rod
3 Rods
Industry and Water Treatment, Ideal for Sampling From Baths, Tanks, Shafts, Open Water Courses, Used with Telescope Sampler
Aluminum
Aluminum
115 to 300 cm (Adjustable)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.