missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Telescopic Rod

Telescopic Rod is a versatile sampling system with exchangeable tools for a wide range of applications.

Brand:  Buerkle™ 5355-0180

52.35 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15376297

  • 57.70 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Different tools can be attached to the telescopic rod
  • The tools (angular beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net) are attached to the telescopic rod by a practical snap-in joint
  • Ideal for sampling from baths, tanks, shafts, open water courses etc
  • Sampling depths down to 6 m can be reached with the adjustable telescopic aluminum rod
  • The telescopic rods cannot be rotated radially, which means even better handling
  • The individual rods have been provided with an end stop, this means that one rod cannot accidentally slip out of another rod
  • Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm. Fits into every car boot!
  • Springs made of stainless steel
Specifications

Specifications

Telescopic Rod
4 Rods
70 to 180 cm
Aluminum
Industry and Water Treatment, Ideal for Sampling From Baths, Tanks, Shafts, Open Water Courses, Used with Telescope Sampler
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers