Learn More
Tellurium, 99+%, (trace metal basis), powder, Thermo Scientific Chemicals
27.30 € - 108.00 €
Chemical Identifiers
CAS | 13494-80-9 |
---|---|
Molecular Formula | Te |
Molecular Weight (g/mol) | 127.60 |
MDL Number | MFCD00134062 |
InChI Key | PORWMNRCUJJQNO-UHFFFAOYSA-N |
Synonym | tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds |
PubChem CID | 6327182 |
ChEBI | CHEBI:30452 |
IUPAC Name | tellurium |
SMILES | [Te] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10603322
|
Thermo Scientific Acros
420020100 |
10 g | Glass bottle |
27.30 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10627591
|
Thermo Scientific Acros
420021000 |
100 g | Glass bottle |
108.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13494-80-9 | |
127.60 | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
6327182 | |
tellurium |
Te | |
MFCD00134062 | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
CHEBI:30452 | |
[Te] |
Specifications
990.0°C | |
13494-80-9 | |
Powder | |
Te | |
MFCD00134062 | |
15, 9263 | |
Solubility in water: insoluble. Other solubilities: soluble in h2so4, hno3 and koh, insoluble in benzene and carbon disulfide | |
[Te] | |
127.60 | |
CHEBI:30452 | |
≥99% | |
Tellurium, 99+% |
450.0°C | |
Black to Gray | |
99+% | |
Te | |
15,298 | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
tellurium | |
6327182 | |
127.6 | |
Trace Metal Basis |
Safety and Handling
GHS H Statement
Harmful if inhaled.
May cause an allergic skin reaction.
May cause long lasting harmful effects to aquatic life.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 236-813-4
missing translation for 'rtecsNumber' : WY2625000
missing translation for 'tsca' : TSCA
RUO – Research Use Only