missing translation for 'onlineSavingsMsg'
Learn More

Tellurium broken ingot, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11396078
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 13494-80-9
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
Synonym tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
PubChem CID 6327182
ChEBI CHEBI:30452
IUPAC Name tellurium
SMILES [Te]

Spezifikation

Quantity 250 g
Percent Purity 99.99%
Odor Odorless
Form Ingot
Assay (metals basis)
Chemical Name or Material Tellurium broken ingot

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'