missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11361629
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tellurium(IV) chloride is used in organic synthesis to prepare Cl-C-C-TeCl3derivatives by reacting with alkenes and aryl tellurium compounds by reacting with electron rich arenes. It is involved in the preparation of diaryl telluride from anisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-07-0
Molecular Formula Cl4Te
Molecular Weight (g/mol) 269.4
MDL Number MFCD00011262
InChI Key SWLJJEFSPJCUBD-UHFFFAOYSA-N
Synonym tellurium tetrachloride, telluric chloride, tellurium chloride, tetrachlorotellurium, tellurium iv chloride, tellurium chloride tecl4 , t-4, unii-dny2r5498h, tellurium chloride, t-4, telluricchloride, tecl4
PubChem CID 61443
SMILES Cl[Te](Cl)(Cl)Cl

Specifications

Boiling Point 380°C
Melting Point 224°C
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Decomposes in water forming TeO2 and HClSoluble in acetonitrile,dimethyl sulfoxide and dimethylformamide. Slightly soluble in ether and benzene. Insoluble in cyclohexane,hexanes,methylene chloride and chloroform.
Formula Weight 269.41
Sensitivity Moisture sensitive
Density 3.01 g/mL
Chemical Name or Material Tellurium(IV) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.