missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) chloride, 99%

Product Code. p-221638 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10702241

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10026-07-0 | Cl4Te | 269.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-07-0
Molecular Formula Cl4Te
Molecular Weight (g/mol) 269.4
MDL Number MFCD00011262
InChI Key SWLJJEFSPJCUBD-UHFFFAOYSA-N
Synonym tellurium tetrachloride, telluric chloride, tellurium chloride, tetrachlorotellurium, tellurium iv chloride, tellurium chloride tecl4 , t-4, unii-dny2r5498h, tellurium chloride, t-4, telluricchloride, tecl4
PubChem CID 61443
SMILES Cl[Te](Cl)(Cl)Cl

Specifications

Boiling Point 380.0°C
Melting Point 224.0°C
Color White to Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 98.5% min. (ex Cl) (Argentometry)
Linear Formula TeCl4
Fieser 09,443; 10,377; 12,457; 16,316
Merck Index 15, 9268
Solubility Information Solubility in water: may decompose. Other solubilities: forming teo2 and hcl, soluble in absolute alcohol and toluene
Formula Weight 269.4
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 3.26
Density 3.2600g/mL
Chemical Name or Material Tellurium(IV) chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Tellurium(IV) chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.