missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) iodide, 99% (metals basis)

Product Code. 11355039
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11355039

Brand: Thermo Scientific Alfa Aesar 087795.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-48-9 | I4Te | 635.218 g/mol

Tellurium(IV) iodide is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-48-9
Molecular Formula I4Te
Molecular Weight (g/mol) 635.218
MDL Number MFCD00049578
InChI Key XCOKHDCPVWVFKS-UHFFFAOYSA-N
Synonym tellurium tetraiodide, tellurium iv iodide, tellurium iodide tei4 , t-4, unii-zb48i033v5, tetraiodo-, e4-tellane, tei4, tellurium iv iodide-te
PubChem CID 82255
SMILES [Te](I)(I)(I)I

Specifications

Boiling Point (decomposition)
Melting Point 280°C
Physical Form Crystalline
Quantity 25 g
UN Number UN2923
Merck Index 14,9127
Solubility Information Decomposes slowly in cold water to TeO2 and HI
Formula Weight 635.22
Percent Purity 99%
Odor Acrid
Sensitivity Moisture sensitive
Density 5.403 g/mL at 15°C
Assay (metals basis)
Chemical Name or Material Tellurium (IV) iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.