Learn More
Tellurium(IV) oxide, tech., Thermo Scientific Chemicals
132.66 € - 498.48 €
Chemical Identifiers
CAS | 7446-07-3 |
---|---|
Molecular Formula | O2Te |
Molecular Weight (g/mol) | 159.60 |
MDL Number | MFCD00011263 |
InChI Key | LAJZODKXOMJMPK-UHFFFAOYSA-N |
Synonym | tellurium iv oxide, tellurium oxide teo2, tellurite teo2, teo2, unii-397e9rke83, oxotellane oxide, telluride oxide, 6-chlorobutyric acid, acmc-1bglq |
PubChem CID | 62638 |
SMILES | O=[Te]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11360769
|
Thermo Scientific Alfa Aesar
010608.18 |
50 g |
132.66 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11350769
|
Thermo Scientific Alfa Aesar
010608.30 |
250 g |
498.48 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as an acousto-optic material and a conditional glass former. The glasses of tellurium oxide have high refractive indices. Owing to the transmission into mid-infrared part of the electromagnetic spectrum, tellurium oxide is of technological interest for optical waveguides. It is also employed in optical fibre amplification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7446-07-3 | |
159.60 | |
LAJZODKXOMJMPK-UHFFFAOYSA-N | |
62638 |
O2Te | |
MFCD00011263 | |
tellurium iv oxide, tellurium oxide teo2, tellurite teo2, teo2, unii-397e9rke83, oxotellane oxide, telluride oxide, 6-chlorobutyric acid, acmc-1bglq | |
O=[Te]=O |
Specifications
733°C | |
Powder | |
MFCD00011263 | |
Practically insoluble in water. Soluble in NaOH solutions and HCl | |
O=[Te]=O | |
159.60 | |
159.6 | |
5.8 |
7446-07-3 | |
O2Te | |
tellurium iv oxide, tellurium oxide teo2, tellurite teo2, teo2, unii-397e9rke83, oxotellane oxide, telluride oxide, 6-chlorobutyric acid, acmc-1bglq | |
LAJZODKXOMJMPK-UHFFFAOYSA-N | |
tellurium dioxide | |
62638 | |
Technical | |
Tellurium(IV) Oxide |
Safety and Handling
P201-P202-P261-P271-P272-P280g-P281-P302+P352-P304+P340-P308+P313-P333+P313-P363-P501c
H317-H332-H360
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 231-193-1
missing translation for 'rtecsNumber' : WY2675000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Reproductive toxicity on or via lactation
- Reproductive toxicity Category 1B
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H362-May cause harm to breast-fed children.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.