missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tellurium powder, -200 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
82.50 € - 1131.00 €
Chemical Identifiers
CAS | 13494-80-9 |
---|---|
Molecular Formula | Te |
Molecular Weight (g/mol) | 127.60 |
MDL Number | MFCD00134062 |
InChI Key | PORWMNRCUJJQNO-UHFFFAOYSA-N |
Synonym | tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds |
PubChem CID | 6327182 |
ChEBI | CHEBI:30452 |
IUPAC Name | tellurium |
SMILES | [Te] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356088
|
Thermo Scientific Alfa Aesar
011075.09 |
10 g |
82.50 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336088
|
Thermo Scientific Alfa Aesar
011075.18 |
50 g |
301.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346088
|
Thermo Scientific Alfa Aesar
011075.30 |
250 g |
1131.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13494-80-9 | |
127.60 | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
6327182 | |
tellurium |
Te | |
MFCD00134062 | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
CHEBI:30452 | |
[Te] |
Specifications
13494-80-9 | |
Powder | |
Te | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
[Te] | |
127.60 | |
CHEBI:30452 | |
Gray powder |
Gray | |
99.999% (Metals basis) | |
MFCD00134062 | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
tellurium | |
6327182 | |
Odorless | |
Tellurium |
Safety and Handling
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 236-813-4
missing translation for 'rtecsNumber' : WY2625000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only