missing translation for 'onlineSavingsMsg'
Learn More
Tellurium powder, -325 mesh, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11345767

Tellurium powder, -325 mesh, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11345767
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11345767

Brand: Thermo Scientific Alfa Aesar 043450.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13494-80-9
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
Synonym tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
PubChem CID 6327182
ChEBI CHEBI:30452
IUPAC Name tellurium
SMILES [Te]

Specifications

UN Number UN3288
Quantity 50 g
Percent Purity 99.99%
Mesh Size −325 Mesh
Odor Odorless
Form Powder
Assay (metals basis)
Chemical Name or Material Tellurium powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY19G002 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number043450.18,043450.09,043465.K7,043465.M6
Lot NumberD18T025 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043450.18,043450.09,043465.K7,043465.M6
Lot NumberC18N06 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043450.18,043450.09,043465.K7,043465.M6
Lot Number60900233 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043450.18,043450.09,043465.K7,043465.M6
Lot NumberF14M24 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043450.18,043450.09,043465.K7,043465.M6
compliance-icons
Product Identifier
  • Tellurium powder
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tellurium powder, -325 mesh, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.