man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Protein Analysis Reagents
Protein Purification
Spin Columns and Gravity-flow Columns
Kinesis™ TELOS Filtration Column
15186763
Print
Kinesis™ TELOS Filtration Column
15mL
Brand:
Kinesis™ 06479-49
View more versions of this product
Product Code.
15186763
328.00 €
/ Pack of 100
Quantity
Estimated Shipment: 02-01-2025
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Certificates
Special Offers
Description
Description
5μm
PTFE
Specifications
Specifications
Capacity (Metric)
15 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers