missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEMED, Approx. 99%, Molecular Biology Reagent, MP Biomedicals™
Click to view available options
Quantity:
25 mL
Specifications
Specifications
Density | 0.775g/mL at 20°C |
Assay Percent Range | Approx. 99% |
Quantity | 25 mL |
Formula Weight | 116.2 |
Grade | Molecular Biology Reagent |
Physical Form | Liquid |
Chemical Name or Material | N,N,N',N'-Tetramethylethylenediamine, FTIR |
Safety and Handling
Product Identifier
- N,N,N',N'-tetramethylethylenediamine, molecular biology reagent
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : N,N,N',N'-tetramethylethylehtylenediamine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction