missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Tempus™ Blood RNA Tube

Tempus Blood RNA Tubes are for stabilization and isolation of total RNA from whole blood for gene expression analysis.

Brand:  Applied Biosystems™ 4342792


Product Code. 12063745

  • 504.00 € / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Tempus Blood RNA Tubes are for stabilization and isolation of total RNA from whole blood for gene expression analysis. Advantages of Tempus Blood RNA Tubes:
• Immediately lyse whole blood cells and stabilize RNA in a single step—no RBC lysis or proteinase K treatments required
• Freeze global gene expression profiles for up to five days at room temperature; greater than seven days at 4°C

How it works
Each Tempus Blood RNA Tube contains 6 mL Stabilizing Reagent. When the blood (3 mL) is drawn into the tube and mixed with this reagent, lysis occurs almost immediately. The stabilizing reagent inactivates cellular RNases and selectively precipitates RNA; genomic DNA (gDNA) while proteins remain in solution. Blood drawn into the Tempus Blood RNA Tube is stable for up to five days at room temperature, or minimally seven days at 4°C.

Accessory product
When the Tempus Blood RNA Tubes are used with the Tempus Spin RNA Isolation Kit (Cat. No. 4380204), RNAs with greater stability and higher yields of RNA are produced.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 Tempus Blood RNA Tubes; room temperature

Whole Blood
Room Temperature
Blood RNA Stabilization Tubes
50 tubes
3 mL of Whole Blood
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tempus Blood RNA Tube

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.

Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.