missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Tenofovir
Reverse transcriptase inhibitor
154.00 € - 525.00 €
Specifications
Purity | 99% |
---|---|
Molecular Formula | (R)-[[2-(6-Amino-9 H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid |
CAS | 147127-20-6 |
Content And Storage | -20°C |
Gene Alias | PMPA, (R)-9-(2-Phosphonoylmethoxypropyl)adenine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18034322
|
Tocris Bioscience™
3666/50 |
50 mg |
525.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18065612
|
Tocris Bioscience™
3666/10 |
10 mg |
154.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selectively inhibits HIV reverse transcriptase (RNA-dependent DNA polymerase). Prevents cytotoxicity in SIV-infected C-8166 cells in vitro (IC50 = 1.5 μM). Antiviral agent.Specifications
99% | |
147127-20-6 | |
PMPA, (R)-9-(2-Phosphonoylmethoxypropyl)adenine | |
M.W. (Observed): 287.21 |
(R)-[[2-(6-Amino-9 H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid | |
-20°C | |
Tenofovir | |
Tenofovir |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title