missing translation for 'onlineSavingsMsg'
Learn More
Tenoxicam, Thermo Scientific Chemicals Product Code.: 15455495

Tenoxicam, Thermo Scientific Chemicals

Product Code. 15455495
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15455495

Brand: Thermo Scientific Alfa Aesar H60837.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59804-37-4
Molecular Formula C13H11N3O4S2
Molecular Weight (g/mol) 337.37
MDL Number MFCD00083502
InChI Key WZWYJBNHTWCXIM-RAXLEYEMSA-N
Synonym tenoxicam, mobiflex, tilcotil, tenoxicamum, liman, tenoxicamum inn-latin, apo-tenoxicam, novo-tenoxicam, tilcotil tn, unii-z1r9n0a399Show MoreShow Less
PubChem CID 54677971
ChEBI CHEBI:32192
IUPAC Name 4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-ylthieno[2,3-e]thiazine-3-carboxamide
SMILES CN1\C(=C(/O)NC2=CC=CC=N2)C(=O)C2=C(C=CS2)S1(=O)=O

Specifications

Quantity 25 g
UN Number UN2811
Formula Weight 337.37
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-thieno[2,3-e]-1,2-thiazine-3-carboxamide 1,1-dioxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81202379 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60837.03,H60837.14,H60837.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tenoxicam, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.