Learn More
Tenuazonsäure-(acetyl-13C2)-Lösung, Honeywell Fluka™
50 μg/mL in acetonitrile, analytical standard, reference material
Brand:  Honeywell Fluka R1323-2ML
Product Code. 15961151
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Specifications
75-05-8, 1486471-66-2
2 mL
0.78 g/cm3
6°C
Liquid
Glass Bottle
Analytical Standard
82°C
Mycotoxins