Learn More
TER-119 Rat anti-Mouse, CoraLite 488, Clone: TER-119, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech CL488-65149-100TESTS
This item is not returnable.
View return policy
Description
The TER-119 antibody from Proteintech is a rat monoclonal antibody generated with day-14 fetal liver cells from a c57bl/6 mouse of mouse TER-119. This antibody recognizes mouse antigen. The TER-119 antibody has been validated for the following applications: FC analysis. This antibody is coralite™488.
Specifications
TER-119 | |
Monoclonal | |
5 μL/Test | |
Rat | |
Primary | |
Mouse |
Flow Cytometry | |
TER-119 | |
CoraLite 488 | |
100 Tests | |
104231 | |
IgG2b |