Learn More
Terazosin hydrochloride dihydrate, MedChemExpress
MedChemExpress Terazosin hydrochloride dihydrate is a quinazoline derivative and a competitive and orally active α1-adrenoceptor antagonist. Terazosin hydrochloride dihydrate works by relaxing blood vessels and the opening of the bladder. Terazosin hydrochloride dihydrate has the potential for benign prostatic hyperplasia (BPH) and high blood pressure treatment.
Brand: Medchem Express HY-B0371A
Description
Terazosin hydrochloride dihydrate is a quinazoline derivative and a competitive and orally active α1-adrenoceptor antagonist. Terazosin hydrochloride dihydrate works by relaxing blood vessels and the opening of the bladder. Terazosin hydrochloride dihydrate has the potential for benign prostatic hyperplasia (BPH) and high blood pressure treatment.Specifications
Terazosin hydrochloride dihydrate | |
White | |
C19H30ClN5O6 | |
DMSO : 31.25 mg/mL (67.95 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (27.18 mM; Need ultrasonic) | |
459.92 | |
99.23% | |
Solid |
70024-40-7 | |
Research | |
100 mg | |
O=C(N1CCN(C2=NC(N)=C3C=C(OC)C(OC)=CC3=N2)CC1)C4OCCC4.O.Cl.O | |
459.92 | |
Research |
Product Suggestions
Customers who viewed this item also viewed