Learn More
Terbinafine hydrochloride
Selective TASK3 activator
Brand:  Tocris Bioscience 6484/50
46.65 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18716188
  • 59.00 € / 50mg
Explore more special offers
Description
Description
Terbinafine hydrochloride is a selective TASK3 activator (pEC50 = 6.2). Exhibits >10-fold selectivity for TASK3 over TREK2, TRESK, THIK1 and TASK2. Also inhibits TWIK1 (pIC50 = 5.69). Also antifungal; inhibits fungal sterol biosynthesis by inhibition of squalene epoxidase.
Specifications
Specifications
78628-80-5
50 mg
327.89
N-[(2E)-6,6-Dimethyl-2-hepten-4-yn-1-yl]-N-methyl-1-naphthalenemethanamine hydrochloride
C21H25N.HCl
98%