Learn More
Terbium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals
90.55 € - 329.64 €
Chemical Identifiers
CAS | 14284-95-8 |
---|---|
Molecular Formula | C15H21O6Tb |
Molecular Weight (g/mol) | 456.25 |
MDL Number | MFCD00013503 |
InChI Key | MAPWZFZLNRMAMO-UHFFFAOYSA-N |
Synonym | terbium iii acetylacetonate, terbium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11 |
PubChem CID | 129878174 |
IUPAC Name | (Z)-4-hydroxypent-3-en-2-one;terbium |
SMILES | [Tb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11369217
|
Thermo Scientific Alfa Aesar
040032.04 |
2 g |
90.55 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11379217
|
Thermo Scientific Alfa Aesar
040032.09 |
10 g |
329.64 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Terbium(III) 2,4-pentanedionate is used as an active pharmaceutical ingredient. It is also used as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14284-95-8 | |
456.25 | |
MAPWZFZLNRMAMO-UHFFFAOYSA-N | |
129878174 | |
[Tb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
C15H21O6Tb | |
MFCD00013503 | |
terbium iii acetylacetonate, terbium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11 | |
(Z)-4-hydroxypent-3-en-2-one;terbium |
Specifications
14284-95-8 | |
C15H21O6Tb | |
terbium iii acetylacetonate, terbium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11 | |
MAPWZFZLNRMAMO-UHFFFAOYSA-N | |
(Z)-4-hydroxypent-3-en-2-one;terbium | |
129878174 | |
99.9% | |
(REO) |
Powder | |
MFCD00013503 | |
Insoluble in water. | |
[Tb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
456.25 | |
456.25 | |
Odorless | |
Terbium(III) 2,4-pentanedionate |
Safety and Handling
missing translation for 'einecsNumber' : 238-195-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.