missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) chloride, anhydrous, 99.9% (REO)

Product Code. p-7051631
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11398818

Brand: Thermo Scientific Alfa Aesar 041678.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10042-88-3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-88-3
Molecular Formula Cl3Tb
Molecular Weight (g/mol) 265.28
MDL Number MFCD00011258
InChI Key GFISHBQNVWAVFU-UHFFFAOYSA-K
IUPAC Name trichloroterbium
SMILES Cl[Tb](Cl)Cl

Specifications

Melting Point 588°C
Physical Form Powder
Quantity 1 g
Merck Index 14,9157
Solubility Information Soluble in water
Formula Weight 265.29
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Terbium(III) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.