missing translation for 'onlineSavingsMsg'
Learn More
Terbium(III) chloride hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10256540

Terbium(III) chloride hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10256540
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10256540

Brand: Thermo Scientific Acros 199610050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13798-24-8
Molecular Formula Cl3H12O6Tb
Molecular Weight (g/mol) 373.37
MDL Number MFCD00149870
InChI Key ULJUVCOAZNLCJZ-UHFFFAOYSA-K
IUPAC Name terbium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]

Specifications

Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 5 g
Assay Percent Range 99.90%
Linear Formula TbCl3·6H2O
Merck Index 14, 9157
Solubility Information Solubility in water: soluble. Other solubilities: readily soluble in alcohol
Formula Weight 373.38
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Terbium(III) chloride hexahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Terbium(III) chloride hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.