Learn More
Terbium(III) chloride hexahydrate, REacton™, 99.999% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 13798-24-8 |
---|---|
Molecular Formula | Cl3H12O6Tb |
Molecular Weight (g/mol) | 373.37 |
MDL Number | MFCD00149870 |
InChI Key | ULJUVCOAZNLCJZ-UHFFFAOYSA-K |
IUPAC Name | terbium(3+) hexahydrate trichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3] |
Description
Terbium(III) chloride hexahydrate is a water soluble source used for the preparation of metallic terbium, which is used in electronic devices and an additive for special alloy namely Terfenol-D. It is used in the preparation of complexes, which are used in the spectrofluorimetric determination of various chemicals in pharmaceutical formulations. Further, it finds use in the preparation of terbium nanotubes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13798-24-8 | |
373.37 | |
ULJUVCOAZNLCJZ-UHFFFAOYSA-K | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3] |
Cl3H12O6Tb | |
MFCD00149870 | |
terbium(3+) hexahydrate trichloride |
Specifications
13798-24-8 | |
Cl3H12O6Tb | |
14,9157 | |
ULJUVCOAZNLCJZ-UHFFFAOYSA-K | |
terbium(3+) hexahydrate trichloride | |
373.38 (265.29 Anhydrous) | |
Reacton™ | |
(metals basis) |
Crystalline | |
MFCD00149870 | |
Very soluble in water. | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3] | |
373.37 | |
99.999% | |
Odorless | |
Terbium (III) chloride hexahydrate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Warning
missing translation for 'einecsNumber' : 233-132-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only