missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11396928
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13813-40-6
MDL Number MFCD00049573

Specifications

Melting Point 946°C
Physical Form Flakes
Quantity 5 g
Formula Weight 539.64
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Terbium(III) iodide, Ultra dry

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'