missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III,IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

281.40 € - 561.80 €

Chemical Identifiers

CAS 12037-01-3
Molecular Formula O7Tb4
Molecular Weight (g/mol) 747.69
MDL Number MFCD00083151
InChI Key UWZVBPNIJCKCMC-UHFFFAOYSA-N
IUPAC Name tetraoxotetraterboxane
SMILES O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326248
SDS View Documents
Thermo Scientific Alfa Aesar
011207.09
10 g
281.40 €
10g
Please to purchase this item. Need a web account? Register with us today!
11316248
SDS View Documents
Thermo Scientific Alfa Aesar
011207.18
50 g
561.80 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Terbium(III,IV) oxide acts as a precursor for the preparation of other terbium compounds. It is used as a redox catalyst in reactions involving oxygen. At high temperatures, it can liberate oxygen. It finds applications in the manufacture of glass, optical and ceramic. Nanoparticles of terbium oxide are used as analytical reagents for the determination of drugs in food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12037-01-3
747.69
UWZVBPNIJCKCMC-UHFFFAOYSA-N
O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O
O7Tb4
MFCD00083151
tetraoxotetraterboxane

Specifications

2340°C
Powder
O7Tb4
Soluble in dilute acids. Insoluble in water.
O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O
747.69
Terbium(III, IV) oxide
12037-01-3
99.9% (REO)
MFCD00083151
UWZVBPNIJCKCMC-UHFFFAOYSA-N
tetraoxotetraterboxane
747.69
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-856-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Terbium(III,IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.