missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III,IV) oxide, REacton™, 99.998% (REO), Thermo Scientific Chemicals

Product Code. 11311448
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It acts as a precursor for the preparation of other terbium compounds. It is used as a redox catalyst in reactions involving oxygen. At high temperatures, it can liberate oxygen. It finds applications in the manufacture of glass, optic and ceramic. Nanoparticles of terbium oxide are used as analytical reagents for the determination of drugs in food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12037-01-3
Molecular Formula O7Tb4
Molecular Weight (g/mol) 747.69
MDL Number MFCD00083151
InChI Key UWZVBPNIJCKCMC-UHFFFAOYSA-N
IUPAC Name tetraoxotetraterboxane
SMILES O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O

Specifications

Melting Point 2340°C
Physical Form Powder
Quantity 5 g
Merck Index 14,9157
Solubility Information Soluble in dilute acids. Insoluble in water.
Formula Weight 747.69
Percent Purity 99.998%
Grade Reacton™
Odor Odorless
Assay (REO)
Chemical Name or Material Terbium(III,IV) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.