missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) nitrate hydrate, 99.9% (REO)

Product Code. 11364879
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11364879

Brand: Thermo Scientific Alfa Aesar 074103.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Terbium(III) nitrate hydrate, H2N3O10Tb, CAS Number-94219-56-4, terbium 3+ hydrate trinitrate, 3no3.tb.h2o, 25g, 99.9% (REO), Hazard Class: 5.1; Packaging Group: III, 233-138-7, 344.94 (anhydrous), H272-H315-H319-H335 | CAS: 94219-56-4 | N3O9Tb | 344.94 g/mol

Terbium(III) nitrate hexahydrate forms a microporous material, which is used for sensor applications due to its luminescent properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94219-56-4
Molecular Formula N3O9Tb
Molecular Weight (g/mol) 344.94
MDL Number MFCD00011257
InChI Key YJVUGDIORBKPLC-UHFFFAOYSA-N
Synonym 3no3.tb.h2o, terbium 3+ hydrate trinitrate
PubChem CID 91886285
IUPAC Name terbium(3+);trinitrate;hydrate
SMILES [Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Wet Crystalline Aggregates
Quantity 25 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
Formula Weight 344.94 (anhydrous)
Chemical Name or Material Terbium(III) nitrate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.