missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terbium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Terbium(III) nitrate hexahydrate forms a microporous material, which is used for sensor applications due to its luminescent properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Wet Crystalline Aggregates |
Quantity | 5 g |
Assay Percent Range | 99.9% (REO) |
Solubility Information | Soluble in water. |
Formula Weight | 344.94 (Anhydrous) |
Chemical Name or Material | Terbium(III) nitrate hydrate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Terbium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction