missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

86.60 € - 1375.00 €

Chemical Identifiers

CAS 148980-31-8
Molecular Formula C3F9O9S3Tb
Molecular Weight (g/mol) 606.115
MDL Number MFCD00192461
InChI Key JLYAVYSVLLIWEK-UHFFFAOYSA-K
Synonym terbium iii trifluoromethanesulfonate, terbium tritriflate, terbium iii triflate, terbium tris trifluoromethanesulfonate, trifluoromethanesulfonic acid terbium iii salt, terbium trifluoromethanesulfonate, terbium triflate, tb otf 3, acmc-1byi6
PubChem CID 11786785
IUPAC Name terbium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tb+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11353868
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040312.04
2 g
86.60 €
2g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11333868
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040312.09
10 g
293.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11343868
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040312.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

It is used as a reagent & chemical intermediates. It is a raw material used in synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

148980-31-8
606.115
JLYAVYSVLLIWEK-UHFFFAOYSA-K
11786785
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tb+3]
C3F9O9S3Tb
MFCD00192461
terbium iii trifluoromethanesulfonate, terbium tritriflate, terbium iii triflate, terbium tris trifluoromethanesulfonate, trifluoromethanesulfonic acid terbium iii salt, terbium trifluoromethanesulfonate, terbium triflate, tb otf 3, acmc-1byi6
terbium(3+);trifluoromethanesulfonate

Specifications

148980-31-8
C3F9O9S3Tb
terbium iii trifluoromethanesulfonate, terbium tritriflate, terbium iii triflate, terbium tris trifluoromethanesulfonate, trifluoromethanesulfonic acid terbium iii salt, terbium trifluoromethanesulfonate, terbium triflate, tb otf 3, acmc-1byi6
JLYAVYSVLLIWEK-UHFFFAOYSA-K
terbium(3+);trifluoromethanesulfonate
11786785
98%
Powder
MFCD00192461
Insoluble in water.
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tb+3]
606.115
606.12
Terbium(III) trifluoromethanesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only