Learn More
Terbium rod, 12.7mm (0.50in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Form: Approx. 1.04g/mm; Cast rod, diameter is approximate
900.48 € - 1422.08 €
Chemical Identifiers
CAS | 7440-27-9 |
---|---|
Molecular Formula | Tb |
Molecular Weight (g/mol) | 158.93 |
MDL Number | MFCD00011256 |
InChI Key | GZCRRIHWUXGPOV-UHFFFAOYSA-N |
Synonym | unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic |
PubChem CID | 23958 |
ChEBI | CHEBI:33376 |
IUPAC Name | terbium |
SMILES | [Tb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11365108
|
Thermo Scientific Alfa Aesar
044049.HF |
10 mm |
900.48 €
10mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11375108
|
Thermo Scientific Alfa Aesar
044049.DH |
25 mm |
1422.08 €
25mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-27-9 | |
158.93 | |
GZCRRIHWUXGPOV-UHFFFAOYSA-N | |
23958 | |
terbium |
Tb | |
MFCD00011256 | |
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendriticShow More | |
CHEBI:33376 | |
[Tb] |
Specifications
7440-27-9 | |
Cast Rod | |
Tb | |
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic | |
[Tb] | |
158.93 | |
CHEBI:33376 | |
Terbium |
Silver to Gray | |
99.9% (Metals basis) | |
MFCD00011256 | |
GZCRRIHWUXGPOV-UHFFFAOYSA-N | |
terbium | |
23958 | |
Silver Gray rod |
Safety and Handling
missing translation for 'einecsNumber' : 231-137-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.