Learn More
Terephthaloyl Chloride, 99+%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 100-20-9 |
---|---|
Molecular Formula | C8H4Cl2O2 |
Molecular Weight (g/mol) | 203.02 |
MDL Number | MFCD00000693 |
InChI Key | LXEJRKJRKIFVNY-UHFFFAOYSA-N |
Synonym | terephthaloyl chloride, terephthaloyl dichloride, 1,4-benzenedicarbonyl dichloride, terephthalic acid dichloride, terephthalic dichloride, p-phthaloyl chloride, terephthalic acid chloride, p-phthalyl dichloride, p-phthaloyl dichloride, terephthalyl dichloride |
PubChem CID | 7488 |
IUPAC Name | benzene-1,4-dicarbonyl chloride |
SMILES | ClC(=O)C1=CC=C(C=C1)C(Cl)=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-20-9 | |
203.02 | |
LXEJRKJRKIFVNY-UHFFFAOYSA-N | |
7488 | |
ClC(=O)C1=CC=C(C=C1)C(Cl)=O |
C8H4Cl2O2 | |
MFCD00000693 | |
terephthaloyl chloride, terephthaloyl dichloride, 1,4-benzenedicarbonyl dichloride, terephthalic acid dichloride, terephthalic dichloride, p-phthaloyl chloride, terephthalic acid chloride, p-phthalyl dichloride, p-phthaloyl dichloride, terephthalyl dichloride | |
benzene-1,4-dicarbonyl chloride |
Specifications
100-20-9 | |
White | |
99+% | |
C6H4(COCl)2 | |
09, 844 | |
LXEJRKJRKIFVNY-UHFFFAOYSA-N | |
benzene-1,4-dicarbonyl chloride | |
7488 | |
≥99.0% | |
Terephthaloyl chloride, 99+% |
79°C to 83°C | |
Authentic | |
C8H4Cl2O2 | |
MFCD00000693 | |
terephthaloyl chloride, terephthaloyl dichloride, 1,4-benzenedicarbonyl dichloride, terephthalic acid dichloride, terephthalic dichloride, p-phthaloyl chloride, terephthalic acid chloride, p-phthalyl dichloride, p-phthaloyl dichloride, terephthalyl dichloride | |
ClC(=O)C1=CC=C(C=C1)C(Cl)=O | |
203.02 | |
203.02 | |
Crystalline Flakes, Crystals or Chunks |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-829-5
missing translation for 'rtecsNumber' : WZ1797000
missing translation for 'tsca' : TSCA
- Terephthaloyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only