Learn More
Abnova™ TERF2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand:  Abnova™ H00007014-T01.100uL
Product Code. 16121322
  • 215.00 € / 100µL
Explore more special offers
Specifications
293T
TERF2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 28.3kDa
Transient overexpression cell lysate was tested with anti-TERF2 antibody (Mfr. No. H00007014-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
BC024890
pCMV-TERF2 full-length
100 μL
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Abnova™ TERF2 293T Cell Transient Overexpression Lysate (Denatured) > 100μL