missing translation for 'onlineSavingsMsg'
Learn More

α-Terpineol, 97+%, Thermo Scientific Chemicals

Product Code. 15214704
Click to view available options
Quantity:
25 g
500 g
2.5 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-55-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001557
InChI Key WUOACPNHFRMFPN-UHFFFAOYSA-N
Synonym alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
PubChem CID 17100
ChEBI CHEBI:22469
IUPAC Name 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
SMILES CC1=CCC(CC1)C(C)(C)O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 31.0°C to 34.0°C
Color Colorless
Density 0.9370g/mL
Boiling Point 214.0°C to 224.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Refractive Index 1.4800 to 1.4830
Quantity 2.5 kg
Beilstein 06,II,66
Merck Index 15,9313
Specific Gravity 0.937
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents , diethyl ether,, ethanol, benzene, propylene glycol
Formula Weight 154.25
Percent Purity 97+%
Physical Form Liquid After Melting
Chemical Name or Material α-Terpineol, 99%
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.